Merck
  • H-point standard addition method for the selective simultaneous determination of nickel and copper using 1-(2-pyridylazo)-2-naphthol in Tween 80 micellar media.
H-point standard addition method for the selective simultaneous determination of nickel and copper using 1-(2-pyridylazo)-2-naphthol in Tween 80 micellar media.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2004-08-06)
Habibollah Eskandari, Yousef Kamali
ABSTRAKT
The H-point standard addition method (HPSAM) has been applied for the simultaneous determination of nickel and copper in trace levels, using 1-(2-pyridylazo)-2-naphthol (PAN) as a chromogenic reagent in aqueous Tween 80 micellar media. Under the optimum condition, the simultaneous determinations of nickel and copper by HPSAM were performed. The absorbances at one pair of wavelengths, 548 and 579 nm, were monitored with the addition of standard solutions of copper. The method is able to accurately determine copper-to-nickel ratios of 15:1 to 1:10 (Wt/Wt). The effects of diverse ions on the determination of nickel and copper to investigate the selectivity of the method were also studied. The recommended procedure was successfully applied to some water and alloy samples.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1-(2-Pyridylazo)-2-naphthol, indicator grade