Merck
Nondaily hormonal contraception: establishing a fit between product characteristics and patient preferences.
The Journal of family practice (2004-11-06)
Jeffrey P Levine
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ethisterone, ≥99% (HPLC)