Merck
Brazilin protects cultured rat hepatocytes from BrCCl3-induced toxicity.
Drug and chemical toxicology (1992-01-01)
C K Moon, K S Park, S G Kim, H S Won, J H Chung
ABSTRAKT
Brazilin, the main constituent of Caesalpinia sappan, is an antioxidative substance that has catechol moiety in its chemical structure. Considering the antioxidant-activity of brazilin, it was expected to have protective effects on the toxicities of radical generating chemicals. The incubation of rat hepatocytes with BrCCl3 resulted in significant increase in lipid peroxidation, leakage of cytoplasmic enzymes and cytoplasmic glutathione depletion. The BrCCl3-induced toxicities on hepatocytes were reduced by the treatment of brazilin. Brazilin has been also proved to have a protective effect on the BrCCl3-induced depression of microsomal calcium sequestration activity. These results indicate that brazilin plays a protective role in BrCCl3-induced hepatocyte injury of the rat.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Bromotrichloromethane, 99%