Merck
  • p-Iodophenol-enhanced luminol chemiluminescent assay applied to discrimination between acute lymphoblastic and minimally differentiated acute myeloid (FAB-M0) or acute megakaryoblastic (FAB-M7) leukemias.
p-Iodophenol-enhanced luminol chemiluminescent assay applied to discrimination between acute lymphoblastic and minimally differentiated acute myeloid (FAB-M0) or acute megakaryoblastic (FAB-M7) leukemias.
The hematology journal : the official journal of the European Haematology Association (2004-12-01)
Miriane da Costa, Valdecir Farias Ximenes, Iguatemy Lourenço Brunetti, Roberto Passetto Falcão, Luiz Marcos da Fonseca
ABSTRAKT
In this report, we propose the application of the p-iodophenol-enhanced luminol chemiluminescent technique to the determination of peroxidase (myeloperoxidase and/or platelet peroxidase) activity in blasts of minimally differentiated acute myeloblastic leukemia (AML-M0) and acute megakaryoblastic leukemia (AML-M7). The frozen blast cells from 29 patients were thawed and submitted to the optimized protocol. All cases of AML-M7 and AML-M I exhibited integrated light emission greater than 73(10(2) mV x s), which was the arbitrary cutoff point set for the discrimination between AML and acute lymphoblastic leukemia (ALL) (mean + 3 x s.d. of ALL samples, n = 10). In addition, five out of seven cases of AML-MO showed results above the cutoff point. This highly sensitive enhanced chemiluminescent technique may be applied to discriminate between ALL and AML-M7 or AML-MI cases, and most AML-M0 cases. It is very simple, cheap and easy to perform compared to other procedures used to measure MPO activity in AML-leukemias including AML-M7 and AML-M0.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
4-Iodophenol, 99%