Merck
[Pyrogallic method for the determination of protein in urine (literature survey)].
Klinicheskaia laboratornaia diagnostika (2004-12-09)
Iu V Kim, A N Shibanov
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Pyrogallol Red, Suitable for use as a complexometric indicator