Merck
[Studies on the constituents from the stems of Spatholobus suberectus].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2004-12-25)
Jun Cheng, Hong Liang, Yuan Wang, Yu-Ying Zhao
ABSTRAKT
To investigate the constituents from the Stems of Spatholobus suberectus. The chemical constituents were isolated by column chromatography and their strutures were identified on base of physical and spectral data. Five compounds were isolated and identified as 2-methoxy-4-(2'-hydroxyethyl)-phenyl-1-O-beta-D-glucopyranoside (1), n-butyl-O-beta-D-fructo pyranoside (2), glycerol-alpha-pentacosanoate (3), betulinic acid (4) and hexacosanoic acid (5). The five compounds were obtained for the frist time from the stems of Spatholobus suberectus.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hexacosanoic acid, ≥95% (capillary GC)
Sigma-Aldrich
Hexacosanoic acid, technical, ≥90% (GC)