Merck
Effects of 4-tert-pentylphenol on the gene expression of P450 11beta-hydroxylase in the gonad of medaka (Oryzias latipes).
Aquatic toxicology (Amsterdam, Netherlands) (2005-01-12)
Hirofumi Yokota, Tatsuo Abe, Makoto Nakai, Hidekazu Murakami, Chizumi Eto, Yoshikuni Yakabe
ABSTRAKT
Alkylphenols including 4-tert-pentylphenol (4-PP) have been shown to alter sexual differentiation in fish due to their estrogenic properties. Medaka (Oryzias latipes) is so sensitive to these substances because morphological sex reversal and testis-ova induction are well developed in the exposed males. However, little work has been done to characterize the molecular effects of estrogenic substances on the morphological and gonadal feminization in male fish. Cytochrome P450 11beta-hydroxylase (P450(11beta)) is a key steroidogenic enzyme in production of 11-ketotestosterone which is the predominant androgen in male fish. In this study, we cloned a cDNA encoding medaka testicular P450(11beta), and then investigated the gene expression of P450(11beta) in the testes of genetically male medaka exposed to 4-PP. The cDNA contains 1740 nucleotides that encode a protein of 543 amino acids, which shares 68.9% and 73.4% homology with testicular P450(11beta)s from Japanese eel (Anguilla japonica) and rainbow trout (Oncorhynchus mykiss), respectively. HeLa cells transfected with an expression vector containing the open reading frame of medaka P450(11beta) cDNA showed 11beta-hydroxylating activity in the presence of exogenous testosterone. Analysis of tissue distribution by RT-PCR showed great abundance of P450(11beta) mRNA in testis. In the partial life-cycle exposure with 4-PP, morphologically sex-reversal was observed in XY medaka exposed to 4-PP concentrations of > or =238 microg/L. Furthermore, exposure to 4-PP completely inhibited P450(11beta) mRNA expression in the gonads of sex-reversed XY fish at 60-day posthatch. These results suggest that xeno-estrogen 4-PP may have inhibitory effects on the synthesis of testicular 11-oxygenated androgens through downregulation of P450(11beta) expression in the genetically male fish.