- Neutral acetohydroxamic acid coordination to a mononuclear Ni(II) center stabilized by an intramolecular hydrogen-bonding interaction.
Neutral acetohydroxamic acid coordination to a mononuclear Ni(II) center stabilized by an intramolecular hydrogen-bonding interaction.
Chemical communications (Cambridge, England) (2005-01-18)
Katarzyna Rudzka, Magdalena M Makowska-Grzyska, Ewa Szajna, Atta M Arif, Lisa M Berreau
PMID15654379
ABSTRAKT
Treatment of a new chelate ligand having both amide- and phenyl-appended pyridyl moieties with Ni(ClO4).6H2O and acetohydroxamic acid in methanol solution results in the production of a novel pseudo-octahedral Ni(II) complex having a neutral acetohydroxamic acid ligand stabilized by a hydrogen-bonding interaction.
MATERIAŁY