Merck
  • Synthesis and biological evaluation with plant cells of new fosmidomycin analogues containing a benzoxazolone or oxazolopyridinone ring.
Synthesis and biological evaluation with plant cells of new fosmidomycin analogues containing a benzoxazolone or oxazolopyridinone ring.
Journal of enzyme inhibition and medicinal chemistry (2005-01-25)
Martine Courtois, Zoia Mincheva, Françoise Andreu, Marc Rideau, Marie-Claude Viaud-Massuard
ABSTRAKT
Fosmidomycin, 3-(N-formyl-N-hydroxyamido) propylphosphonic acid sodium salt, is an efficient inhibitor of 1-deoxy-D-xylulose-5-phosphate (DOXP) reductoisomerase, the second enzyme of the 2C-methyl-D-erythritol-4-phosphate (MEP) pathway notably present in Plasmodium species. We have synthesized a new series of analogues of fosmidomycin, containing a benzoxazolone, benzoxazolethione or oxazolopyridinone ring. As the MEP pathway is involved in the biosynthesis of all isoprenoids, accumulation of ajmalicine in Catharanthus roseus cells was chosen as a marker of monoterpenoid indole alkaloid (MIA) production. None of the twelve studied phosphonic esters 3 and phosphonic acids 4 affected periwinkle cell growth, but some of them (3c, 3e, 3g and 3h) showed a significant inhibition of ajmalicine accumulation: 45-85% at 125 microM. Surprisingly, this effect disappeared by conversion of 3c and 3g into the corresponding acids 4c and 4g, respectively.