Merck
[FTIR investigation of new polymer solid electrolytes].
Guang pu xue yu guang pu fen xi = Guang pu (2005-03-16)
Shu-ting Yang, Hong-jun Chen, Hong-yu Dong, Jun-hua Jia, Zhao-xia Cao
ABSTRAKT
The conductivity of the porous polymer solid electrolyte blended with PVDF and PMMA, which was made by a micro-wave hot-cross-linking method, reached 2.05 x 10(-3) S x cm(-1) at room temperature. The polymer solid electrolyte was analyzed and investigated by FTIR. The results show that the PVDF, PMMA and LiClO4 in the polymer solid electrolyte were not simply blended, but certain kind of effect existed which was strengthened only when the polymer solid electrolyte came into being.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Lithium perchlorate, purum p.a., ≥98.0% (calc. based on dry substance, T), powder
Sigma-Aldrich
Lithium perchlorate, ACS reagent, ≥95.0%
Sigma-Aldrich
Lithium perchlorate, 99.99% trace metals basis
Sigma-Aldrich
Lithium perchlorate, battery grade, dry, 99.99% trace metals basis