Merck
3-phenylpropionate catabolism and the Escherichia coli oxidative stress response.
Research in microbiology (2005-04-06)
Evelyne Turlin, Odile Sismeiro, Jean Pierre Le Caer, Valérie Labas, Antoine Danchin, Francis Biville
ABSTRAKT
Cells have devised a variety of protection systems against the toxic effects of dioxygen. Dioxygenases are part of this defence mechanism. In Escherichia coli, the positive regulator HcaR, a member of the LysR family of regulators, controls expression of the neighbouring genes, hcaA1, hcaA2, hcaC, hcaB and hcaD, coding for the 3-phenylpropionate dioxygenase complex and 3-phenylpropionate-2',3'-dihydrodiol dehydrogenase, that oxidizes 3-phenylpropionate to 3-(2,3-dihydroxyphenyl) propionate. Differences between expression of hcaR and expression of its target, hcaA, suggest that HcaR is involved in control of other cellular processes or that other regulatory proteins modulate hcaA expression. Protein expression profiling was used to identify other HcaR targets. Two-dimensional gel electrophoresis was used to compare the proteomes of wild-type E. coli and strains in which hcaR was disrupted. Several polypeptides whose production was up- or downregulated in the hcaR mutant were involved in the oxidative stress response. Subsequent experiments demonstrated that hcaR disruption was involved in regulation of genes involved in the oxidative stress response. Modification of the stress response also occurred in an hcaA1A2CD mutant strain. Using gel retardation, the HcaR binding site was estimated to be located about -70 to -55 bp upstream of the hcaA transcription start site. The expression of hcaR was repressed in the absence of oxygen by the ArcA/ArcB two-component system.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hydrocinnamic acid, 99%
Sigma-Aldrich
3-Phenylpropionic acid, 99%, FG