Merck
A psbA mutation in Kochia scoparia (L) Schrad from railroad rights-of-way with resistance to diuron, tebuthiuron and metribuzin.
Pest management science (2005-06-14)
Lemma W Mengistu, Michael J Christoffers, Rodney G Lym
ABSTRAKT
Kochia [Kochia scoparia (L) Schrad] has become resistant to many herbicides used in cropland and railroad rights-of-way in North Dakota and Minnesota. Kochia scoparia plants that had survived annual treatments with diuron and tebuthiuron were sampled along railroad rights-of-way in North Dakota and Minnesota. The samples were screened in the greenhouse for resistance to diuron, tebuthiuron, metribuzin and bromoxynil from 0.5x to 32x the recommended use rates. A resistant K scoparia accession (MN-3R) was confirmed with resistance up to 16-fold higher than recommended use rates for tebuthiuron and diuron and up to 4-fold higher for metribuzin. However, the resistant K scoparia accession was susceptible to bromoxynil even at 50% of the recommended use rate. The herbicide binding region of the psbA gene fragment of eight resistant (R) and seven susceptible (S) K scoparia accessions was PCR-amplified and sequenced for detection of mutations. The psbA gene of four R K scoparia accessions was mutated at residue 219 with substitution of isoleucine for valine (GenBank accession number AY251265). The seven S K scoparia accession sequences were wild-type at this residue (GenBank accession number AY251266). The other four R accessions sequences showed a previously known triazine R mutation with substitution of glycine for serine at residue 264. All 15 K scoparia accessions were wild-type at all other psbA residues within the region analyzed. Resistance to diuron, tebuthiuron and metribuzin among the railroad rights-of-way K scoparia is probably due to the mutation at residue 219 of the psbA gene in some plants, but due to the previously reported Ser(264)Gly substitution in other plants. Target-site resistance associated with a change of valine to isoleucine at residue 219 of the psbA target-site in weeds has previously been reported for Poa annua L selected in diuron-treated grass seed fields, and for Amaranthus powelli S Wats selected in linuron-treated carrot fields. This is the first report of the mutation in herbicide-resistant K scoparia.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Tebuthiuron, PESTANAL®, analytical standard