Merck
Rate-limiting biotransformation of triamterene is mediated by CYP1A2.
International journal of clinical pharmacology and therapeutics (2005-07-23)
U Fuhr, S Kober, M Zaigler, E Mutschler, H Spahn-Langguth
ABSTRAKT
Triamterene (TA), a potassium-sparing diuretic, is extensively metabolized by hydroxylation in 4'-position and subsequent conjugation by cytosolic sulfotransferases. To identify the cytochrome P450 enzyme(s) catalyzing hydroxylation of triamterene (the rate-limiting step in the formation of the sulfate ester (STA)), in vitro incubation studies were performed with human liver microsomes. Initial rates of TA hydroxylation (0 - 300 microM) were determined during a ten-minute-incubation period with liver microsomes of two donors. The role of individual CYP enzymes was determined by pre-incubation with selective inhibitors/alternative substrates. Vice versa, the effect of TA (0 - 500 microM) on 3-demethylation of caffeine (0 - 1,000 microM) was assessed. Metabolite concentrations were estimated by reversed-phase HPLC methods. TA Km values without inhibitors were 60 and 142 microM, Vmax was 177 and 220 pmol/min/mg protein, respectively. Mean inhibitor induced changes of 4'-hydroxy-TA formation were as follows: Furafylline 25 microM (CYP1A2), complete inhibition (-100%); omeprazole 250 microM (CYP1A2 inhibitor/CYP2C 19 substrate), -30%; coumarin 25 microM (CYP2A6), -11%; quinidine 25 microM (CYP2D6), -9%; ketoconazole 25 microM (CYP3A), -18%; and erythromycin 250 microM (CYP3A), -8%. In the reverse inhibition studies, TA competitively inhibited caffeine 3-demethylation with Ki values of 65 and 111 microM, respectively. 4'-hydroxylation of TA in humans appears to be mediated exclusively by CYP1A2. Inhibition or induction of CYP1A2 will change the time course of both TA and its active phase-II metabolite. The net pharmacodynamic effect of such changes is difficult to predict and needs to be evaluated in clinical studies.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Furafylline, ≥98% (HPLC)