Merck
Use of chlorite to improve HPLC detection of pyridoxal 5'-phosphate.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2005-08-02)
Karen L Ericson, J Dennis Mahuren, Yvonne M Zubovic, Stephen P Coburn
ABSTRAKT
The sensitivity of fluorescent detection of the biologically active form of Vitamin B-6, pyridoxal 5'-phosphate (PLP), in biological samples has been improved approximately four-fold by adopting chlorite as a post-column derivatization reagent (instead of bisulfite) in high-performance liquid chromatography (HPLC) separation. Chlorite oxidizes PLP to the more fluorescent 4-pyridoxic acid 5'-phosphate, and avoids the toxicity and heating of the cyanide procedure. Detection of another major metabolite, 4-pyridoxic acid (4-PA), is not effected. Detection of pyridoxal (PL) is slightly lowered due to eluting at a lower pH.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
4-Pyridoxic acid, ≥98%