Merck
Enhancement of cisplatin sensitivity of cisplatin-resistant human cervical carcinoma cells by bryostatin 1.
Clinical cancer research : an official journal of the American Association for Cancer Research (2005-09-17)
Sanghamitra Mohanty, Jie Huang, Alakananda Basu
ABSTRAKT
Bryostatin 1, a unique protein kinase C (PKC) activator, is already in the clinical trials. An understanding of complex regulation of PKC by bryostatin 1 is essential for effective use of bryostatin 1 in the clinic. We have previously shown that the ability of bryostatin 1 to enhance cisplatin sensitivity correlated with its ability to down-regulate PKCdelta in HeLa cells. We have investigated how bryostatin 1 influences PKCdelta regulation in cisplatin-resistant HeLa (HeLa/CP) cells, and if bryostatin 1 could be used to reverse cisplatin resistance. Phorbol 12,13-dibutyrate (PDBu), bryostatin 1, and small interfering RNA were used to manipulate PKC level/activation status. Cell death was monitored by 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) assay, Annexin V dye-binding assay, and analysis of hypodiploid peak in a flow cytometer. Bryostatin 1 elicited a biphasic concentration response on PKCdelta down-regulation and cisplatin-induced cell death in HeLa/CP cells; the maximum effect was achieved with 1 nmol/L bryostatin 1. Down-regulation of PKCalpha increased with increasing concentrations of bryostatin 1. PDBu induced down-regulation of PKCalpha in HeLa and HeLa/CP cells but it had little effect on PKCdelta down-regulation in HeLa/CP cells. However, both PDBu and bryostatin 1 enhanced the sensitivity of HeLa/CP cells to cisplatin. Knockdown of PKCdelta by small interfering RNA inhibited cisplatin-induced apoptosis but knockdown of PKCalpha enhanced cisplatin-induced cell death. These results suggest that although PKCdelta acts as a proapoptotic protein, full-length PKCdelta may inhibit cisplatin-induced cell death. Thus, persistent activation/down-regulation of PKCdelta by bryostatin 1 was associated with cisplatin sensitization. Furthermore, PKCalpha acts as an antiapoptotic protein and down-regulation of PKCalpha by PDBu was associated with cellular sensitization to cisplatin.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
(−)-Indolactam V, ≥96% (HPLC)