Merck
Palladium-catalyzed allylation of acidic and less nucleophilic anilines using allylic alcohols directly.
Chemical & pharmaceutical bulletin (2005-10-06)
Yi-Chun Hsu, Kim-Hong Gan, Shyh-Chyun Yang
ABSTRAKT
The direct activation of C-O bonds in allylic alcohols by palladium complexes has been accelerated by carrying out the reactions in the presence of titanium(IV) isoproxide and 4 A molecular sieves. The acidic and less nucleophilic anilines such as diphenylamine, phenothiazine, 4-cyanoaniline, and nitroanilines are efficiently allylated under palladium catalysis using allylic alcohols as allylating reagents.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Titanium(IV) isopropoxide, 99.999% trace metals basis
Sigma-Aldrich
Titanium(IV) isopropoxide, 97%
Sigma-Aldrich
Titanium(IV) isopropoxide, ≥97.0%
Sigma-Aldrich
Titanium(IV) isopropoxide, packaged for use in deposition systems