Merck
The immunological activities of bacterial peptidoglycans.
Annual review of microbiology (1980-01-01)
D E Stewart-Tull
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Peptidoglycan from Staphylococcus aureus, cell wall component