Merck
Effects of systemic administration of beta-casomorphin-5 on learning and memory in mice.
European journal of pharmacology (2005-12-20)
Minoru Sakaguchi, Makoto Koseki, Masanori Wakamatsu, Eiko Matsumura
ABSTRAKT
The effects of systemic administration of bovine beta-casomorphin-5 (Tyr-Pro-Phe-Pro-Gly), a mu-opioid receptor agonist derived from milk beta-casein, on spontaneous alternation behavior in the Y-maze (spatial short-term memory) and step-down-type passive avoidance response (non-spatial long-term memory) were investigated in mice. Intraperitoneal (i.p.) administration of beta-casomorphin-5 (0.1-20 mg/kg) did not have a significant effect on either spontaneous alternation behavior or passive avoidance response. However, a low dose (1 mg/kg, i.p.) of beta-casomorphin-5 improved scopolamine (1 mg/kg, s.c.)-induced impairment of spontaneous alternation behavior and passive avoidance response. Pretreatment with intracerebroventricular injections of beta-funaltrexamine (a mu-opioid receptor antagonist, 0.1 microg/mouse) and naloxonazine (a mu(1)-opioid antagonist, 5 microg/mouse), which did not improve scopolamine-induced impairment, prevented the ameliorating effect of beta-casomorphin-5 on scopolamine-induced impairment of passive avoidance response. These results indicated that systemic administration of a low dose (1 mg/kg, i.p.) of beta-casomorphin-5 improves the disturbance of learning and memory resulting from cholinergic dysfunction through central mediation involving mu(1)-opioid receptors.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
β-Casomorphin Fragment 1-5 hydrochloride, ≥97% (HPLC)