Merck
Isolation of the active compound in Mauria heterophylla, a Peruvian plant with antibacterial activity.
Phytotherapy research : PTR (2006-01-31)
Tatsuya Mori, Cecilia Chang, Dora Maurtua, Gerald B Hammond
ABSTRAKT
A fraction from the ethanol extract of the Peruvian medicinal plant Mauria heterophylla (Anacardiaceae) showed antibacterial activity against Escherichia coli 35992, Staphylococcus aureus 20213 and Pseudomonas aeruginosa 15442. Further fractionation led to the isolation and characterization of ethyl gallate as the antibacterial active compound.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ethyl gallate, antioxidant, ≥96.0% (HPLC)