- [Studies on chemical constituents of Patrinia villosa].
[Studies on chemical constituents of Patrinia villosa].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2006-03-31)
Jin-Yong Peng, Guo-Rong Fan, Yu-Tian Wu
PMID16570799
ABSTRAKT
To investigate the chemical constituents of Patrinia villosa. The chemical constituents were isolated by silica gel column chromatography and semi-preparative high-performance liquid chromatography, and identified by physicochemical properties and spectral analysis (MS, 1H-NMR and 13C-NMR). Seven compounds were isolated from ethyl acetate and n-butanol extract and identified as: 5-hydroxyl-7, 3', 4'-trimethoxy flavone (I), 5-hydroxyl-7, 4'-dimethoxy flavone (II), luteolin (III), quercetin (IV), isoorientin (V), isovitexin (VI) and 8-C glucosylprunetin (VII). Compounds I , II, III, V, VI and VIII were obtained from the plant of genus Patrinia for the first time, compound IV was separated from P. villosa for the first time.