Merck
Emergency management of seizures in children.
Lancet (London, England) (2006-05-16)
Monisha Goyal, Max Wiznitzer
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Paraldehyde, ≥97.0% (GC)
Sigma-Aldrich
Paraldehyde, Arxada quality, ≥99.5% (GC)