Merck
Tricyclic quinoxalinediones, aza-kynurenic acids, and indole-2-carboxylic acids as in vivo active NMDA-glycine antagonists.
Current topics in medicinal chemistry (2006-05-25)
Ryu Nagata, Seiji Katayama, Ken-Ichi Ohtani, Hiroyasu Tanaka, Kozo Shimago
ABSTRAKT
This review article describes the development of in vivo active antagonists for the glycine binding site of the N-Methyl-D-Aspartate (NMDA) receptor. There were several difficulties in identifying a class of antagonists with in vivo efficacy and only a few compounds succeeded in emerging with activity in vivo. A series of tricyclic quinoxalinediones was highly potent glycine antagonists in vitro and the derivatives having a zwitterionic moiety including SM-18400 indeed showed in vivo activity. Similarly, tricyclic indole-2-carboxylic acids having a zwitterionic moiety such as SM-31900 were also active in vivo. In fact, SM-18400 and SM-31900 exhibited efficacy in several animal stroke models using intravenous infusion protocols. The practical syntheses of SM-18400 and SM-31900 as well as the novel synthesis of moderately active glycine antagonists, tricyclic azakynurenic acids, were also developed.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Indole-2-carboxylic acid, 98%