Merck
Physicochemical properties and antitumor activities of water-soluble native and sulfated hyperbranched mushroom polysaccharides.
Carbohydrate research (2006-06-30)
Yongzhen Tao, Lina Zhang, Peter C K Cheung
ABSTRAKT
A water-soluble hyperbranched beta-glucan, coded as TM3b, extracted from sclerotia of an edible fungus (Pleurotus tuber-regium) was fractioned into eight fractions coded as F1-F8 by a nonsolvent addition method. Five fractions were treated with chlorosulfonic acid at 35 degrees C to synthesize successfully sulfated derivatives coded as S-F2, S-F3, S-F4, S-F5, and S-F8 with degree of substitution of 0.28-0.54. The 13C NMR results of these sulfated beta-glucans indicated that while the C-6 position was fully substituted, C-2, C-3, and C-4 were only partially substituted by the sulfate groups. The weight-average molecular weights (Mw) and intrinsic viscosities ([eta]) of the native and sulfated TM3b fractions were determined using multi-angle laser light scattering and viscometry in 0.15M aq NaCl at 25 degrees C, respectively. The dependences of [eta] on Mw for TM3b and sulfated TM3b were found to be [eta]=0.18Mw(0.28+/-0.03) (Mw range from 3.30 x 10(4) to 3.90 x 10(7)) and [eta]=2.24 x 10(-2)Mw(0.52+/-0.06) (Mw range from 3.24 x 10(4) to 3.15 x 10(5)) in 0.15M aq NaCl at 25 degrees C, respectively. It revealed that both the native TM3b and its sulfated derivatives exist in a spherical chain conformation in 0.15M aq NaCl. Furthermore, the native and sulfated TM3b fractions showed potent antitumor activities in vivo and in vitro. The sulfated derivatives exhibited relatively higher in vitro antitumor activity against human hepatic cancer cell line HepG2 than the native TM3b. Water solubility and introduction of sulfate groups were the main factors in enhancing the antitumor activities.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Chlorosulfonic acid, 99%