Merck
[Forecasting parameters of acute chemical toxicity according to in vitro conformation changes of proteins].
Meditsina truda i promyshlennaia ekologiia (2006-07-04)
A S Luk'ianov, T K Semina, A M Korolev
ABSTRAKT
The authors analyze experimental data on in vitro effects induced by chemicals that were used throughout MEIC toxicologic studies in ovalbumin and acetylcholinesterase of human RBC. Influence on proteins is compared to acute toxicity caused by the chemicals in humans and various cell lines. The conclusion is that the method is prospective for screening of acute chemical toxicity signs in humans.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Potassium cyanate-15N, ≥98 atom % 15N, ≥95% (CP)
Sigma-Aldrich
Potassium cyanate, 96%