Merck
Degradation and reconstruction of moenomycin A and derivatives: dissecting the function of the isoprenoid chain.
Journal of the American Chemical Society (2006-10-26)
Masaatsu Adachi, Yi Zhang, Catherine Leimkuhler, Binyuan Sun, John V LaTour, Daniel E Kahne
ABSTRAKT
Moenomycin A is the only known natural product that inhibits peptidoglycan biosynthesis by binding the bacterial transglycosylases. We describe a degradation/reconstruction route to manipulate the reducing end of moenomycin A. A comparison of the biological and enzyme inhibitory activity of moenomycin A and an analogue containing a nerol lipid in place of the natural C25 lipid chain provides insight into the role of the moenocinol unit. Our results show that a lipid chain having ten carbons in moenocinol is sufficient for enzyme inhibition, but a longer chain is required for biological acitivity, apparently because the molecule must partition into biological membranes to reach its target in bacterial cells.