Merck
Antidepressant-like effects of sarsasapogenin from Anemarrhena asphodeloides BUNGE (Liliaceae).
Biological & pharmaceutical bulletin (2006-11-02)
Li-Xiang Ren, Yi-Fan Luo, Xia Li, Dai-Ying Zuo, Ying-Liang Wu
ABSTRAKT
The aim of this study was to investigate the effects of sarsasapogenin from Anemarrhena asphodeloides BUNGE (Liliaceae) on the forced swimming test, and the central noradrenergic, dopaminergic and serotonergic activities in mice. Our results showed that sarsasapogenin treatment at 12.5, 25 and 50 mg/kg (p.o.) for 14 d significantly reduced the duration of immobility in the forced swimming test. These doses that affected the immobile response did not affect locomotor activity. In addition, the neurochemical assays showed that sarsasapogenin produced a marked increase of noradrenaline and serotonin levels at 50 mg/kg in both the hypothalamus and the hippocampus. Moreover, sarsasapogenin showed a monoamine oxidase inhibitory activity in the mouse brain. These findings suggest that the antidepressant activity of sarsasapogenin may involve the central monoaminergic neurotransmitter systems.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sarsasapogenin, ≥98%