Merck
NMR spectra of some reduced symmetry peripheral fused-ring-substituted phthalocyanines: density functional calculations.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2006-11-14)
Xianxi Zhang, Zhongqiang Liu, Nagao Kobayashi, Jianzhuang Jiang
ABSTRAKT
Chemical shifts of some reduced symmetry peripheral fused-ring-substituted phthalocyanines, namely Zn3B1N, Zncis2B2N, Zntrans2B2N, Zn1B3N and Zn3B0N, have been calculated at density functional B3LYP level using the gauge-independent atomic orbital (GIAO) method. The geometries were optimized using the 6-31G(d) basis set and the following NMR calculations were performed using 6-31G(d) and 6-311G(d,p) basis sets, respectively. The calculated NMR shielding tensors and chemical shifts are compared with previous experimental results. The chemical shifts are assigned according to the calculated data and satisfying results are obtained. The NMR shielding tensor simulation of Zn3B0N has been raised as a significant theoretical topic.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Tetramethylsilane, analytical standard, suitable for NMR (spectroscopy), ACS reagent
Sigma-Aldrich
Tetramethylsilane, electronic grade, ≥99.99% trace metals basis
Sigma-Aldrich
Tetramethylsilane, ≥99.0% (GC)
Sigma-Aldrich
Tetramethylsilane, NMR grade, ACS reagent, ≥99.9%