Merck
ACAT inhibition of alkamides identified in the fruits of Piper nigrum.
Phytochemistry (2006-12-26)
Mun-Chual Rho, Seung Woong Lee, Hye Ran Park, Jung-Ho Choi, Ji Yun Kang, Koanhoi Kim, Hyun Sun Lee, Young Kook Kim
ABSTRAKT
In this study, via a bioactivity-guided fractionation of MeOH extracts of the fruits of Piper nigrum, alkamide (5) and five previously-identified alkamides were isolated. Their structures were elucidated via spectroscopic analysis ((1)H, (13)C NMR and ESI-MS), as follows: retrofractamide A (1), pipercide (2), piperchabamide D (3), pellitorin (4), dehydroretrofractamide C (5) and dehydropipernonaline (6). The IC(50) values determined for the compounds were 24.5 (1), 3.7 (2), 13.5 (3), 40.5 (4), 60 (5) and 90 microM (6), according to the results of an ACAT enzyme assay system using rat liver microsomes. These compounds all inhibited cholesterol esterification in HepG2 cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Cholesteryl oleate, ≥98% (HPLC; detection at 205 nm)