Merck
Atropisomers of meso-conjugated uracyl porphyrin derivatives and their assembling structures.
Organic letters (2006-12-29)
Satoshi Arai, Daisuke Niwa, Hiroyuki Nishide, Shinji Takeoka
ABSTRAKT
[reaction: see text] We have synthesized a 5,15 meso-substituted methyluracyl porphyrin derivative bearing 6-methyluracyl units directly at the meso positions. The atropisomerization was regulated by steric replusion between the methyl substituents. When the atropisomers were mixed with alkylated melamine as a complementary hydrogen-bonding unit, the hydrogen-bonded assemblies were analyzed by diffusion-ordered spectroscopy (DOSY) in solution, which clarified that the alphabeta isomer formed a face-to-face dimer, whereas the alphabeta isomer took a zigzag structure.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Thioanisole, ReagentPlus®, ≥99%