Merck
  • Regio- and stereoselective anomeric esterification of glucopyranose 1,2-diols and a facile preparation of 2-O-acetylated glucopyranosyl trichloroacetimidates from the corresponding 1,2-diols.
Regio- and stereoselective anomeric esterification of glucopyranose 1,2-diols and a facile preparation of 2-O-acetylated glucopyranosyl trichloroacetimidates from the corresponding 1,2-diols.
Carbohydrate research (2007-02-16)
Jianjun Zhang, Xiaomei Liang, Daoquan Wang, Fanzuo Kong
ABSTRAKT
A highly regio- and stereoselective anomeric esterification of 3-O-allyl (or benzyl, or benzoyl)-4,6-O-isopropylidene-alpha,beta-d-glucopyranose with acetyl chloride, or allyl chloroformate, or ethyl chloroformate gave the corresponding 2-OH, 1-beta-acetates or -carbonates in excellent yields. The 2-OH, 1-beta-acetates were readily converted to the corresponding 2-O-acetylated glucopyranosyl trichloroacetimidates by reaction with trichloroacetonitrile via base promoted acetyl migration, while the 2-OH, 1-beta-carbonates were good glycosyl acceptors for the synthesis of (1-->2)-linked oligosaccharides.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acetyl chloride, reagent grade, 98%
Sigma-Aldrich
Acetyl chloride, reagent grade, 98%
Sigma-Aldrich
Ethyl chloroformate, 97%
Sigma-Aldrich
Ethyl chloroformate, purum, ≥98.0% (GC)
Sigma-Aldrich
Trichloroacetonitrile, 98%
Sigma-Aldrich
Allyl chloroformate, 97%
Sigma-Aldrich
Acetyl chloride, puriss. p.a., ≥99.0% (T)