Merck
In the developing rat hippocampus a tonic GABAA-mediated conductance selectively enhances the glutamatergic drive of principal cells.
The Journal of physiology (2007-02-24)
Ivan Marchionni, Azar Omrani, Enrico Cherubini
ABSTRAKT
In the adult hippocampus, two different forms of GABA(A) receptor-mediated inhibition have been identified: phasic and tonic. The first is due to the activation of GABA(A) receptors facing the presynaptic releasing sites, whereas the second is due to the activation of receptors localized away from the synapses. Because of their high affinity and low desensitization rate, extrasynaptic receptors are persistently able to sense low concentrations of GABA. Here we show that, early in postnatal life, between postnatal day (P) 2 and P6, CA1 and CA3 pyramidal cells but not stratum radiatum interneurons, express a tonic GABA(A)-mediated conductance. Block of the neuronal GABA transporter GAT-1 slightly enhanced the persistent GABA conductance in principal cells but not in GABAergic interneurons. However, in adulthood, a tonic GABA(A)-mediated conductance could be revealed in stratum radiatum interneurons, indicating that the ability of these cells to sense ambient GABA levels is developmentally regulated. Pharmacological analysis of the tonic conductance in principal cells demonstrated the involvement of beta2/beta 3, alpha 5 and gamma 2 GABA(A) receptor subunits. Removal of the tonic depolarizing action of GABA with picrotoxin, reduced the excitability and the glutamatergic drive of principal cells but did not modify the excitability of stratum radiatum interneurons. The increased cell excitability and synaptic activity following the activation of extrasynaptic GABA(A) receptors by ambient GABA would facilitate the induction of giant depolarizing potentials.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
NO-711 hydrochloride, ≥98% (HPLC)