Merck
Theoretical study of the structures and vibrational spectra of the hydrogen-bonded systems of 4-cyanophenol with N-bases.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2007-03-06)
Yordanka Dimitrova, Jordan A Tsenov
ABSTRAKT
The structural and vibrational features of the hydrogen bonded complexes of 1,5,7-triazabicyclo [4.4.0] dec-5-ene (TBD) with one and two 4-CNPhOH molecules have been studied extensively by ab initio SCF/6-31G(d,p) and BLYP calculations with various basis sets: 6-31G(d,p), 6-31+G(d,p) and 6-31++G(d,p). Full geometry optimization was made for the complexes studied. The nature of the hydrogen bonding and the influence of the hydrogen bonding on the structural and vibrational characteristics of the monomers have been investigated. The corrected values of the dissociation energy for the hydrogen-bonded complexes have been calculated in order to estimate their stability. The calculated values of the dissociation energy per phenol molecule indicate that the complex: TBD: 4-CNPhOH (1:1) is more stable than the complex: TBD: 4-CNPhOH (1:2). The changes in the structural and vibrational characteristics upon hydrogen bonding depend on the strength of the hydrogen bonds. In agreement with the experiment, the calculations show that the complexation between TBD and 4-CNPhOH leads to considerably changes in the vibrational characteristics of the stretching O-H vibration. The vibrational frequency of the O-H stretching vibration is shifted to lower wave numbers upon hydrogen bonding. The predicted frequency shifts Deltanu(O-H) for the complexes--TBD: 4-CNPhOH (1:1) and TBD: 4-CNPhOH (1:2) are in the range from -190 cm(-1) to -586 cm(-1). In the same time the IR intensity of the O-H stretching vibration increases dramatically in the hydrogen-bonded complexes.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
4-Cyanophenol, 95%