Merck
Method for measuring the self-assembly of alkanethiols on gold at femtomolar concentrations.
Langmuir : the ACS journal of surfaces and colloids (2007-05-08)
Kishan Rijal, Raj Mutharasan
ABSTRAKT
We describe a cantilever-based method for measuring the self-assembly of alkanethiols on a gold surface in a flow system that permits easy step changes in concentration and acquire a continuous in situ measure of the resulting chemisorption through the change in resonance frequency. A gold-coated (2.2 mm2), piezoelectric-excited, millimeter-sized cantilever (PEMC) sensor was exposed to 1-hexadecanethiol (HDT) in ethanol at concentrations ranging from 1 fM to 1 mM, sequentially and separately. A high-order flexural mode at approximately 850 kHz was monitored during the self-assembly. The resonance frequency decreases as a result of increased mass as chemisorption occurs on the surface. We show for the first time that the chemisorption of HDT at 1 fM is readily measurable and gave a response of 220 +/- 13 Hz (n = 4). At higher concentrations (10 and 100 fM; 1, 10, and 100 pM; 1, 10, and 100 nM; 1 microM; and 1 mM), the responses were proportionately, but nonlinearly, higher. At high concentrations (1 mM), the responses to C4, C8, C11, C16, and C18 alkanethiols were linearly proportional and were complete in approximately 25 min. We report for the first time that, once the Au surface is equilibrated at 1 pM, further chemisorption at a lower HDT concentration does not take place, even though over 99% of surface adsorption sites are available. At 1 fM, the overall chemisorption rate did not increase with a 2-fold increase in the HDT flow rate, suggesting that chemisorption at 1 fM is not transport-limited. The measured overall chemisorption rate constant at 1 fM was more rapid than 0.1 min-1.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1-Hexadecanethiol, 99%
Sigma-Aldrich
1-Hexadecanethiol, ≥95.0% (GC)