Merck
  • Synthesis of N-(5,7-diamino-3-phenyl-quinoxalin-2-yl)-3,4,5-substituted anilines and N-[4[(5,7-diamino-3-phenylquinoxalin-2-yl)amino]benzoyl]-l-glutamic acid diethyl ester: evaluation of in vitro anti-cancer and anti-folate activities.
Synthesis of N-(5,7-diamino-3-phenyl-quinoxalin-2-yl)-3,4,5-substituted anilines and N-[4[(5,7-diamino-3-phenylquinoxalin-2-yl)amino]benzoyl]-l-glutamic acid diethyl ester: evaluation of in vitro anti-cancer and anti-folate activities.
European journal of medicinal chemistry (2007-05-29)
Paola Corona, Mario Loriga, M Paola Costi, Stefania Ferrari, Giuseppe Paglietti
ABSTRAKT
Several diamino quinoxalines were designed, synthesized and evaluated as anti-tumor agents. Two compounds showed the most potent cytotoxic activities against the leukemia CCRF-CEM cell line (GI(50)<0.01microM) and the ovarian cancer cell line OVCAR-4 (GI(50)=0.03microM), respectively, with comparable/better activities than Methotrexate (MTX). Docking calculations of the complexes of hDHFR with the most active compounds identified the binding mode of the described molecules with respect to MTX.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
L-Glutamic acid diethyl ester hydrochloride, 97%
CCRF-CEM, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 85112105, human blood, Lymphoblast