Merck
Runoff of pesticides from rice fields in the Ile de Camargue (Rhône river delta, France): field study and modeling.
Environmental pollution (Barking, Essex : 1987) (2007-06-15)
Laetitia Comoretto, Bruno Arfib, Romain Talva, Philippe Chauvelon, Marc Pichaud, Serge Chiron, Patrick Höhener
ABSTRAKT
A field study on the runoff of pesticides was conducted during the cultivation period in 2004 on a hydraulically isolated rice farm of 120 ha surface with one central water outlet. Four pesticides were studied: Alphamethrin, MCPA, Oxadiazon, and Pretilachlor. Alphamethrin concentrations in runoff never exceeded 0.001 microg L(-1). The three other pesticides were found in concentrations between 5.2 and 28.2 microg L(-1) in the runoff water shortly after the application and decreased thereafter. The data for MCPA compared reasonably well with predictions by an analytical runoff model, accounting for volatilization, degradation, leaching to groundwater, and sorption to soil. The runoff model estimated that runoff accounted for as much as 18-42% of mass loss for MCPA. Less runoff is observed and predicted for Oxadiazon and Pretilachlor. It was concluded that runoff from rice paddies carries important loads of dissolved pesticides to the wetlands in the Ile de Camargue, and that the model can be used to predict this runoff.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Pretilachlor, PESTANAL®, analytical standard
Supelco
Oxadiazon, PESTANAL®, analytical standard