Merck
Spectrofluorimetric determination of 5-hydroxytryptamine using chlorosulphonylthenoyltrifluoroacetone-europium probe.
Luminescence : the journal of biological and chemical luminescence (2007-07-05)
Xiaojing Zhu, Xiaolei Wang, Chongqiu Jiang
ABSTRAKT
A new spectrofluorimetric method was developed for the determination of trace amounts of 5-hydroxytryptamine (5-HT). Using chlorosulphonylthenoyltrifluoroacetone (CTTA)-europium (Eu(3+)) ion as a fluorescent probe in a buffer solution at pH 11.0, 5-HT can remarkably enhance the fluorescence intensity of the CTTA-Eu(3+) complex at lambda = 612 nm; the enhanced fluorescence intensity of Eu(3+) is proportional to the concentration of 5-HT. Optimum conditions for the determination of 5-HT were also investigated. The linear range and detection limit for the determination of 5-HT were 1.0 x 10(-7)-1.2 x 10(-5) mol/L and 8.5 x 10(-8) mol/L, respectively. This method is simple, practical and relatively free of interference from coexisting substances, and can be applied to assess 5-HT in urine samples.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2-Thenoyltrifluoroacetone, 99%