Merck
Role of insecticides in reducing thrips injury to plants and incidence of tomato spotted wilt virus in Virginia market-type peanut.
Journal of economic entomology (2007-09-14)
D Ames Herbert, S Malone, S Aref, R L Brandenburg, D L Jordan, B M Royals, P D Johnson
ABSTRAKT
Tomato spotted wilt virus (family Bunyaviridae, genus Tospovirus, TSWV), transmitted by many thrips species, is a devastating pathogen of peanut, Arachis hypogaea L. TSWV has become a serious problem in the Virginia/Carolina peanut-growing region of the United States. During 2002, TSWV was present in 47% of the North Carolina hectarage and caused a 5% yield reduction in Virginia. Factors influencing levels of TSWV in runner market-type peanut cultivars, which are primarily grown in Alabama, Flordia, Georgia, and Texas, have been integrated into an advisory to help those peanut growers reduce losses. An advisory based on the southeast runner market-type version is currently under development for virginia market-type peanut cultivars that are grown primarily in the Virginia/ Carolina region. A version based on preliminary field experiments was released in 2003. One factor used in both advisories relates to insecticide use to reduce the vector populations and disease incidence. This research elucidated the influence of insecticides on thrips populations, thrips plant injury, incidence of TSWV, and pod yield in virginia market-type peanut. Eight field trials from 2003 to 2005 were conducted at two locations. In-furrow application of aldicarb and phorate resulted in significant levels of thrips control, significant reductions in thrips injury to seedlings, reduced incidence of TSWV, and significant increases in pod yield. Foliar application of acephate after aldicarb or phorate applied in the seed furrow further reduced thrips plant injury and incidence of TSWV and improved yield. These findings will be used to improve the current virginia market-type TSWV advisory.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Phorate, PESTANAL®, analytical standard