Merck
  • FRET-based direct and continuous monitoring of human fucosyltransferases activity: an efficient synthesis of versatile GDP-L-fucose derivatives from abundant D-galactose.
FRET-based direct and continuous monitoring of human fucosyltransferases activity: an efficient synthesis of versatile GDP-L-fucose derivatives from abundant D-galactose.
Chemistry (Weinheim an der Bergstrasse, Germany) (2007-10-12)
Takahiro Maeda, Shin-Ichiro Nishimura
ABSTRAKT
We have developed a facile and versatile protocol for the continuous monitoring of human fucosyltransferases activity by using fluorescence energy resonance transfer (FRET), and have explored the feasibility of its use in an inhibitor screening assay. A convenient sugar nucleotide with a fluorogenic probe, 6-deoxy-6-N-(2-naphalene-2-yl-acetamide)-beta-L-galactopyranos-1-yl-guanosine 5'-diphosphate disodium salt (1), was efficiently synthesized from naturally abundant D-galactopyranose via a key intermediate, 6-azide-1,2,3,4-tetra-O-benzoyl-6-deoxy-beta-L-galactopyranose (10). It was demonstrated that the combined use of the glycosyl donor 1 and a dansylated acceptor substrate, sialyl-alpha2,3-LacNAc derivative (2) allowed us to carry out highly sensitive, direct, and continuous in vitro monitoring of the generation of sialyl Lewis X (SLe x), which is catalyzed by human alpha-1,3-fucosyltransferase VI (FUT-VI). A kinetic analysis revealed that compound 1 was an excellent donor substrate (KM=0.94 microM and Vmax=0.14 microM min(-1)) for detecting human FUT-VI activity. To the best of our knowledge, this synthetic fluorogenic probe is the most sensitive and selective donor substrate for FUT-VI among all of the known GDP-Fuc analogues, including the parent GDP-Fuc. When a dansylated asparagine-linked glycopeptide 20, which is derived from egg yolk was employed as an alternate acceptor substrate, a FRET-based assay with compound 1 could be used to directly monitor the alpha1,6-fucosylation at the reducing terminal GlcNAc residue by human FUT-VIII (KM=175 microM and Vmax=0.06 microM/ min); this indicates that the present method might become a general protocol for the characterization of various mammalian fucosyltransferases in the presence of designated fluorogenic acceptor substrates. The present protocol revealed that compound 23, which was obtained by a 1,3-dipolar cycloaddition between the disodium salt 16 and 1-ethynyl-naphthalene exhibits highly potent inhibitory effects against the FUT-VI-mediated sialyl Lewis X synthesis (IC50=5.4 microM).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Guanosine 5′-diphospho-β-L-fucose sodium salt, ≥85%