Merck
  • Synthesis of new unsymmetrical 4,5-dihydroxy-2-imidazolidinones. Dynamic NMR spectroscopic study of the prototropic tautomerism in 1-(2-benzimidazolyl)-3-phenyl-4,5-dihydroxy-2-imidazolidinone.
Synthesis of new unsymmetrical 4,5-dihydroxy-2-imidazolidinones. Dynamic NMR spectroscopic study of the prototropic tautomerism in 1-(2-benzimidazolyl)-3-phenyl-4,5-dihydroxy-2-imidazolidinone.
Molecules (Basel, Switzerland) (2007-11-01)
Mehdi Ghandi, Abolfazl Olyaei, Farshid Salimi
ABSTRAKT
The acid-catalyzed cyclocondensation in refluxing acetonitrile of aqueous glyoxal with N-heteroaryl-N'-phenylureas 4a-f (heteroaryl = 2-thiazolyl, 2-pyrimidinyl,2-pyrazinyl, 2-pyridinyl, 3-pyridinyl and 2-benzimidazolyl) led to the formation of the corresponding 1-heteroaryl-3-phenyl-4,5-dihydroxy-2-imidazolidinones 5a-f. All the products were characterized by elemental and spectroscopic analyses. The free-energy barrier (Delta G not equal) for prototropic tautomerism in 1-(2-benzimidazolyl)-3-phenyl-4,5-dihydroxy-2-imidazolidinone (5f) was determined by dynamic NMR studies to be 81 +/- 2 KJ mol(-1).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2-Imidazolidone, 96%