- Fragrance material review on trans-2-hexenyl salicylate.
Fragrance material review on trans-2-hexenyl salicylate.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2007-11-23)
A Lapczynski, D McGinty, L Jones, C S Letizia, A M Api
PMID18031887
ABSTRAKT
A toxicologic and dermatologic review of trans-2-hexenyl salicylate when used as a fragrance ingredient is presented.