Merck
PCL-PEG-based nanoparticles grafted with GRGDS peptide: preparation and surface analysis by XPS.
Biomacromolecules (2007-11-28)
Vincent Pourcelle, Sabrina Devouge, Marie Garinot, Véronique Préat, Jacqueline Marchand-Brynaert
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Gly-Arg-Gly-Asp-Ser, ≥97% (HPLC)