Merck
Opposite signs of capacitive microsensor signals upon exposure to the enantiomers of methyl propionate compounds.
Angewandte Chemie (International ed. in English) (2007-12-22)
Petra Kurzawski, Anja Bogdanski, Volker Schurig, Reinhard Wimmer, Andreas Hierlemann
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Methyl DL-lactate, ≥97.0% (GC)
Sigma-Aldrich
(+)-Methyl D-lactate, 98%, optical purity ee: 96% (GLC)
Sigma-Aldrich
(−)-Methyl L-lactate, 98%, optical purity ee: 97% (GLC)