Merck
Small-molecule reagents for cellular pull-down experiments.
Bioconjugate chemistry (2008-01-16)
Xiang Wang, Brandon S Imber, Stuart L Schreiber
ABSTRAKT
Affinity purification of interacting proteins from cellular extracts is a powerful technique for identifying the cellular targets of small molecules. Affinity matrix-based small-molecule reagents are usually prepared by conjugating small molecules of interest to a solid support such as agarose. This protocol describes an efficient and robust method to immobilize small molecules containing a primary alcohol, a common functional group in small molecules, especially in small molecules prepared using diversity-oriented synthesis. This method comprises one element of a systematic approach to the target identification problem in chemical biology.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
CDI, ≥97.0% (T)
Sigma-Aldrich
CDI, reagent grade