Merck
Multifocal electroretinogram in rhodopsin P347L transgenic pigs.
Investigative ophthalmology & visual science (2008-01-29)
Yiu-fai Ng, Henry H L Chan, Patrick H W Chu, Chi-ho To, Brian C Gilger, Robert M Petters, Fulton Wong
ABSTRAKT
Neural ectopic rewiring in retinal degeneration such as retinitis pigmentosa (RP) may form functional synapses between cones and rod bipolar cells that cause atypical signal processing. In this study, the multifocal electroretinograms (mfERGs) of a large animal model of RP, the rhodopsin P347L transgenic (Tg) pig, were measured to examine the sources and nature of altered signal processing. mfERG responses from a 6-week-old Tg pig were recorded before and after sequential application of tetrodotoxin (TTX), N-methyl-D-aspartate (NMDA), 2-amino-4-phosphonobutyric acid (APB), and cis-2,3-piperidinedicarboylic acid (PDA), to identify contributions to the retinal signal from inner retinal neurons, the ON-pathway, the OFF-pathway, and photoreceptors. The mfERG response contributions from different retinal components of in the Tg eyes were estimated and compared with control data from eyes of age-matched wild-type (WT) pigs. There was a prominent difference in the estimates of the inner retinal response and ON-bipolar cell pathway contribution between the Tg and WT mfERG responses. In particular, the early components of the inner retinal contribution were obviously altered in the Tg mfERG. The inner retinal components at approximately 24 and 40 ms appeared to be inverted. Differences in the estimates of OFF-bipolar cell pathway contributions were minimal. There was no change in cone cell responses in the Tg mfERG. In Tg retinas, ectopic synapses formed between cones and rod bipolar cells probably altered signal processing of the ON-bipolar cell pathway. In response to the altered visual signal input from the outer retina, signal processing in inner retinal neurons was also modified.