- A phase separable polycarbonate polymerization catalyst.
A phase separable polycarbonate polymerization catalyst.
Chemical communications (Cambridge, England) (2008-02-20)
Chayanant Hongfa, Jianhua Tian, Jeremy Andreatta, Donald J Darensbourg, David E Bergbreiter
PMID18283354
ABSTRAKT
Polyisobutylene is shown to be a nonpolar phase tag that separates a highly colored salen Cr(III) complex from products but is otherwise kinetically similar to a low molecular weight salen Cr(III) complex in polycarbonate formation by Cr(III)-catalyzed copolymerization of CO2 and cyclohexene oxide.
MATERIAŁY