Merck
  • Temporal decrease of trivalent chromium concentration in a standardized algal culture medium: experimental results and implications for toxicity evaluation.
Temporal decrease of trivalent chromium concentration in a standardized algal culture medium: experimental results and implications for toxicity evaluation.
Bulletin of environmental contamination and toxicology (2008-03-18)
Davide A L Vignati, Mamadou L Beye, Janusz Dominik, Anna O Klingemann, Montserrat Filella, Andrzej Bobrowski, Benoît J Ferrari
ABSTRAKT
The fate of two trivalent chromium salts (nitrate and chloride) in ISO algal culture medium was followed over 72 h; i.e., the typical duration of algal toxicity tests. Fifty percent of the initial Cr spikes was lost from the solutions by 24 h, with losses up to 90% after 72 h. Monitoring of the temporal variability of Cr(III) concentrations in algal culture media appears necessary to better characterize the toxicity of trivalent chromium to algae.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Potassium chromate, ≥99.95% trace metals basis
Sigma-Aldrich
Potassium chromate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chromate tetrahydrate, 99%
Sigma-Aldrich
Sodium chromate, 98%