Merck
  • Pharmaceutical analysis by supercritical fluid chromatography: Optimization of the mobile phase composition on a 2-ethylpyridine column.
Pharmaceutical analysis by supercritical fluid chromatography: Optimization of the mobile phase composition on a 2-ethylpyridine column.
Journal of separation science (2008-04-04)
Claudio Brunelli, Yining Zhao, Melissa-Hanna Brown, Pat Sandra
ABSTRAKT
The separation of neutral, acidic, and basic pharmaceuticals with diverse physicochemical properties by packed column supercritical fluid chromatography (pSFC) on a 2-ethylpyridine column (25 cmx4.6 mm id, 3 mum particles) is presented. The optimization strategy involved separations at 100% methanol (MeOH) and at 50% MeOH/50% ACN while keeping the peak symmetry additives formic acid (FA) and isopropylamine (IPA) at constant levels of 0.25% v/v. By plotting the adjusted retention times as a function of the MeOH/ACN ratio, an optimal modifier ratio composition of 65% MeOH/35% ACN was found. The total set of 26 neutral, acidic, and basic pharmaceuticals was analyzed and the optimal composition experimentally verified. This mobile phase composition is currently used in pharmaceutical method development and open-access generic screening environments.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Isopropylamine, ≥97.0% (GC)
Sigma-Aldrich
Isopropylamine, ≥99.5%
Supelco
Isopropylamine, anhydrous, analytical standard
Sigma-Aldrich
2-Ethylpyridine, 97%