Merck
  • Reverse micellar microextraction for rapid analysis of thiol-containing peptides and amino acids by atmospheric-pressure matrix-assisted laser desorption/ionization ion trap and matrix-assisted laser desorption/ionization time-of-flight mass spectrometry.
Reverse micellar microextraction for rapid analysis of thiol-containing peptides and amino acids by atmospheric-pressure matrix-assisted laser desorption/ionization ion trap and matrix-assisted laser desorption/ionization time-of-flight mass spectrometry.
Rapid communications in mass spectrometry : RCM (2008-04-09)
Kavita Agrawal, Hui-Fen Wu, Kamlesh Shrivas
ABSTRAKT
Simple, rapid and inexpensive one-step reverse micellar microextraction (RMME) procedures were combined with matrix-assisted laser desorption/ionization mass spectrometry (MALDI MS) for the determination of thiol-containing peptides and amino acids. In this investigation, a thiol-containing peptide (HW6) was chosen as model compound to understand the mechanism of RMME. The electrostatic interactions between the thiol-containing peptide and reverse micelles were proposed to be reason for the transfer of analytes from the aqueous phase to the organic phase. Reverse micelles were formed by the cationic surfactant, methyltrioctylammonium chloride (MTOAC). The best extraction efficiency of HW6 was obtained under the following conditions: pH 11.0, ionic strength 5.0 mM of KCl and micelle concentration 7.0 mM of MTOAC. The limits of detection (LODs) obtained for HW6 in water, urine and plasma samples were 0.15, 0.19 and 0.28 microM, respectively, with relative standard deviation (RSD) values in the range +/-8.8-10.5%. The sensitivity obtained in water by the present method was 45-fold higher than that of the conventional use of atmospheric-pressure (AP)-MALDI MS. Furthermore, the applicability of the proposed approach was extended for the determination of thiol-containing amino acids in sample solutions by using MALDI time-of-flight (TOF) MS.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Methyltrioctylammonium chloride, ≥97.0% (AT)