Merck
  • A survey of primary tooth pulp therapy as taught in US dental schools and practiced by diplomates of the American Board Of Pediatric Dentistry.
A survey of primary tooth pulp therapy as taught in US dental schools and practiced by diplomates of the American Board Of Pediatric Dentistry.
Pediatric dentistry (2008-04-12)
Bryan Dunston, James A Coll
ABSTRAKT
The purpose of this study was to repeat a 1997 survey of current pulp therapy practice. The directors of dental school predoctoral pediatric dentistry programs (N=56) and board certified pediatric dentists (N=1200) were surveyed in 2005. More dental schools (83%) taught indirect pulp therapy (IPT) compared to 1997. Significantly more used glass ionomer for IPT with most dental schools and diplomates not re-entering a tooth after IPT. Over 30% of schools and diplomates do direct pulp cops using glass ionomer. For pulpotomy, diluted formocresol usage decreased in dental schools (54%) while ferric sulfate significantly increased (24%) and full strength remained at 22%. Shorter placement of pulpotomy medication was noted and ZOE alone the preferred base. Pulpectomy was advocated by 85% of 2005 schools and diplomates with ZOE filler use decreasing while iodoform/calcium hydroxide filler use increasing. More pediatric dentists are using glass ionomer for IPT and direct pulp capping, and there was a trend away from the use of 1:5 diluted formocresol with more using ferric sulfate for pulpotomy. For pulpectomy, most use ZOE but iodoform pastes and calcium hydroxide have increased in usage since 1997 Disagreements continue concerning when to use certain pulp therapies and some directors and diplomates did not follow the AAPD guidelines.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Iodoform, 99%