Merck
Total synthesis of siomycin A: construction of synthetic segments.
Chemistry, an Asian journal (2008-05-09)
Tomonori Mori, Shuhei Higashibayashi, Taiji Goto, Mitsunori Kohno, Yukiko Satouchi, Kazuyuki Shinko, Kengo Suzuki, Shunya Suzuki, Hiraku Tohmiya, Kimiko Hashimoto, Masaya Nakata
ABSTRAKT
The five practical segments for the total synthesis of siomycin A, that is, the dehydropiperidine segment A (5), the pentapeptide segment B (3), the dihydroquinoline segment C (6), and the beta-phenylselenoalanine dipeptide segments D (7) and E (4), were synthesized. Segment A (5) was constructed by the coupling of the azomethine ylide and the chiral sulfinimine, followed by the stereoselective reduction of the six-membered imine function. Segment B (3) was synthesized by the phenylselenylation of the beta-lactone, stereoselective vinylzinc addition to the chiral sulfinimine, and oxazoline-thioamide conversion. Segment C (6) was prepared by the one-pot olefination of the tetrahydroquinoline N-oxide using triflic anhydride and triethylamine, stereoselective reduction of the methyl ketone function, and regioselective Yb(OTf)(3)-catalyzed epoxide opening by the amino group. Segments D (7) and E (4) were synthesized by coupling of the properly protected beta-phenylselenoalanines.